Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sdz-8
Entrez Gene ID
183834
Gene Name
SKN-1 Dependent Zygotic transcript
Gene Aliases
CELE_C55A6.5
Chromosome Location
Chr.V: 11515082 - 11517767 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073520.3 NP_505921.1 5 - 6 891 117

View Full Product Details