Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
H12C20.7
Entrez Gene ID
179587
Gene Name
Uncharacterized protein
Gene Aliases
CELE_H12C20.7
Chromosome Location
Chr.V: 11566804 - 11572990 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383409.1 13 - 14 4690 66

View Full Product Details