Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-68
Entrez Gene ID
179589
Gene Name
protein-coding
Gene Aliases
CELE_H12C20.3
Chromosome Location
Chr.V: 11573440 - 11575839 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380769.2 NP_001366648.1 4 - 5 672 107

View Full Product Details