Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-88
Entrez Gene ID
185689
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_F43A11.1
Chromosome Location
Chr.V: 11583512 - 11585115 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073538.2 NP_505939.2 5 - 6 673 178

View Full Product Details