Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
daf-11
Entrez Gene ID
179605
Gene Name
protein-coding
Gene Aliases
CELE_B0240.3
Chromosome Location
Chr.V: 11718102 - 11726071 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313082.3 NP_001300011.1 7 - 8 1074 100
NM_073559.8 NP_505960.3 12 - 13 2096 100
NR_132532.1 11 - 12 1733 100

View Full Product Details