Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
daf-11
Entrez Gene ID
179605
Gene Name
protein-coding
Gene Aliases
CELE_B0240.3
Chromosome Location
Chr.V: 11718102 - 11726071 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313082.3 NP_001300011.1 5 - 6 696 162
NM_073559.8 NP_505960.3 10 - 11 1718 162

View Full Product Details