Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vab-8
Entrez Gene ID
179675
Gene Name
protein-coding
Gene Aliases
CELE_K12F2.2
Chromosome Location
Chr.V: 12213327 - 12225938 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073662.6 NP_506063.2 1 - 2 72 97

View Full Product Details