Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
frm-7
Entrez Gene ID
179687
Gene Name
FERM domain-containing protein
Gene Aliases
CELE_C51F7.1
Chromosome Location
Chr.V: 12302484 - 12306978 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073684.5 NP_506085.2 4 - 5 705 92

View Full Product Details