Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lin-25
Entrez Gene ID
179732
Gene Name
Protein lin-25
Gene Aliases
CELE_F56H9.5
Chromosome Location
Chr.V: 12649925 - 12658719 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073761.7 NP_506162.1 12 - 13 3191 121
GenBank mRNA U20168.1 - 12 - 13 3213 121

View Full Product Details