Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-25
Entrez Gene ID
179732
Gene Name
Protein lin-25
Gene Aliases
CELE_F56H9.5
Chromosome Location
Chr.V: 12649925 - 12658719 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073761.7 NP_506162.1 4 - 5 364 146
GenBank mRNA U20168.1 - 4 - 5 386 146

View Full Product Details