Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-25
Entrez Gene ID
179732
Gene Name
Protein lin-25
Gene Aliases
CELE_F56H9.5
Chromosome Location
Chr.V: 12649925 - 12658719 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073761.7 NP_506162.1 8 - 9 2358 101
GenBank mRNA U20168.1 - 8 - 9 2380 101

View Full Product Details