Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scc-3
Entrez Gene ID
179749
Gene Name
Cohesin subunit scc-3
Gene Aliases
CELE_F18E2.3
Chromosome Location
Chr.V: 12763938 - 12767933 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073792.5 NP_506193.1 4 - 5 1882 130

View Full Product Details