Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
inft-2
Entrez Gene ID
179786
Gene Name
FH2 domain-containing protein
Gene Aliases
CELE_F15B9.4
Chromosome Location
Chr.V: 13002316 - 13006987 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073852.8 NP_506253.4 7 - 8 2143 90

View Full Product Details