Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srsx-19
Entrez Gene ID
188760
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_T22G5.4
Chromosome Location
Chr.V: 13887658 - 13889662 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074041.4 NP_506442.1 1 - 2 112 70

View Full Product Details