Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-34A1
Entrez Gene ID
188399
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_T10H4.10
Chromosome Location
Chr.V: 15285556 - 15287956 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074386.2 NP_506787.1 4 - 5 931 118

View Full Product Details