Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-88
Entrez Gene ID
187709
Gene Name
Seven TM Receptor
Gene Aliases
CELE_R08H2.2
Chromosome Location
Chr.V: 15382439 - 15383839 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074420.1 NP_506821.1 5 - 6 769 113

View Full Product Details