Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyb-3
Entrez Gene ID
180040
Gene Name
protein-coding
Gene Aliases
CELE_T06E6.2
Chromosome Location
Chr.V: 15393906 - 15397351 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028941.4 NP_001024112.2 2 - 3 156 86
NM_074424.9 NP_506825.4 3 - 4 1670 86
GenBank mRNA U21282.1 - 2 - 3 159 86

View Full Product Details