Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-33D1
Entrez Gene ID
187025
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_K05D4.4
Chromosome Location
Chr.V: 16173746 - 16175857 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074675.4 NP_507076.1 4 - 5 892 72

View Full Product Details