Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-37
Entrez Gene ID
190828
Gene Name
C-type LECtin
Gene Aliases
CELE_Y102A5B.3
Chromosome Location
Chr.V: 16850394 - 16852102 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074854.5 NP_507255.2 4 - 5 440 100

View Full Product Details