Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srz-94
Entrez Gene ID
186078
Gene Name
Serpentine Receptor, class Z
Gene Aliases
CELE_F49H6.11
Chromosome Location
Chr.V: 17032812 - 17035950 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074913.2 NP_507314.1 1 - 2 145 144

View Full Product Details