Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srj-26
Entrez Gene ID
191272
Gene Name
Serpentine Receptor, class J
Gene Aliases
CELE_ZK262.10
Chromosome Location
Chr.V: 18421097 - 18423477 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075203.1 NP_507604.1 3 - 4 362 75

View Full Product Details