Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- lbp-1
- Entrez Gene ID
-
180616
- Gene Name
- Fatty acid-binding protein homolog 1
- Gene Aliases
-
CELE_F40F4.3
- Chromosome Location
-
Chr.X: 3259087 - 3274756 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076156.7 |
NP_508557.1 |
1 - 2 |
378 |
60 |
- Gene Symbol
- lbp-2
- Entrez Gene ID
-
180617
- Gene Name
- Fatty acid-binding protein homolog 2
- Gene Aliases
-
CELE_F40F4.2
- Chromosome Location
-
Chr.X: 3258976 - 3261470 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076157.9 |
NP_508558.1 |
1 - 2 |
489 |
60 |
View Full Product Details