Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ckc-1
Entrez Gene ID
188971
Gene Name
protein-coding
Gene Aliases
CELE_T27A10.3
Chromosome Location
Chr.X: 3576608 - 3578834 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029758.3 NP_001024929.1 5 - 6 967 106

View Full Product Details