Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lst-2
Entrez Gene ID
180712
Gene Name
Lateral signaling target protein 2
Gene Aliases
CELE_R160.7
Chromosome Location
Chr.X: 4378004 - 4383366 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076355.6 NP_508756.2 1 - 2 106 78

View Full Product Details