Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ptr-4
Entrez Gene ID
180876
Gene Name
SSD domain-containing protein
Gene Aliases
CELE_C45B2.7
Chromosome Location
Chr.X: 6060754 - 6065986 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076612.7 NP_509013.2 7 - 8 1489 89

View Full Product Details