Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hsp-43
Entrez Gene ID
180895
Gene Name
protein-coding
Gene Aliases
CELE_C14F11.5
Chromosome Location
Chr.X: 6233148 - 6236386 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129635.6 NP_001123107.2 1 - 2 155 67
NM_076644.7 NP_509045.1 2 - 3 341 67

View Full Product Details