Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lact-1
Entrez Gene ID
185882
Gene Name
Beta-lactamase domain-containing protein
Gene Aliases
CELE_F46H5.8
Chromosome Location
Chr.X: 7258116 - 7260377 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076820.3 NP_509221.2 6 - 7 691 116

View Full Product Details