Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-27
Entrez Gene ID
181124
Gene Name
Troponin I 2
Gene Aliases
CELE_ZK721.2
Chromosome Location
Chr.X: 8789450 - 8790621 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077087.8 NP_509488.2 2 - 3 430 94

View Full Product Details