Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ubc-21
Entrez Gene ID
182321
Gene Name
Probable ubiquitin-conjugating enzyme E2 21
Gene Aliases
CELE_C06E2.3
Chromosome Location
Chr.X: 8869625 - 8871901 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077101.4 NP_509502.3 3 - 4 343 87

View Full Product Details