Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sto-6
Entrez Gene ID
190619
Gene Name
PHB domain-containing protein
Gene Aliases
CELE_Y71H9A.2
Chromosome Location
Chr.X: 11632407 - 11636533 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077542.7 NP_509943.1 2 - 3 181 86

View Full Product Details