Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
odr-7
Entrez Gene ID
181403
Gene Name
Nuclear hormone receptor family member odr-7
Gene Aliases
CELE_T18D3.2
Chromosome Location
Chr.X: 12458979 - 12461657 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077688.4 NP_510089.1 1 - 2 353 66
GenBank mRNA U16708.1 - 1 - 2 360 66

View Full Product Details