Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxb-26
Entrez Gene ID
186508
Gene Name
FBA_2 domain-containing protein
Gene Aliases
CELE_F58E1.3
Chromosome Location
Chr.II: 1682518 - 1683655 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061638.2 NP_494039.1 1 - 2 103 119

View Full Product Details