Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sre-36
Entrez Gene ID
3564932
Gene Name
Serpentine Receptor, class E (epsilon)
Gene Aliases
CELE_F13A7.13
Chromosome Location
Chr.V: 16399688 - 16401625 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028623.1 NP_001023794.1 3 - 4 623 86

View Full Product Details