Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- sre-36
- Entrez Gene ID
-
3564932
- Gene Name
- Serpentine Receptor, class E (epsilon)
- Gene Aliases
-
CELE_F13A7.13
- Chromosome Location
-
Chr.V: 16399688 - 16401625 on Build WBcel235
- Gene Symbol
- F13A7.14
- Entrez Gene ID
-
6418722
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_F13A7.14
- Chromosome Location
-
Chr.V: 16401272 - 16405348 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383478.1 |
|
1 - 2 |
131 |
94 |
View Full Product Details