Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srz-61
Entrez Gene ID
186935
Gene Name
Serpentine Receptor, class Z
Gene Aliases
CELE_K03D3.4
Chromosome Location
Chr.IV: 16321166 - 16323230 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070541.3 NP_502942.2 2 - 3 268 154

View Full Product Details