Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srz-67
Entrez Gene ID
3565127
Gene Name
Serpentine Receptor, class Z
Gene Aliases
CELE_C17F4.10
Chromosome Location
Chr.II: 3235956 - 3237138 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026794.1 NP_001021965.1 4 - 5 715 126

View Full Product Details