Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-42
Entrez Gene ID
3565737
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_Y54G11A.15
Chromosome Location
Chr.II: 14287618 - 14289068 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027301.3 NP_001022472.1 1 - 2 326 92

View Full Product Details