Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srz-63
Entrez Gene ID
3565730
Gene Name
Serpentine Receptor, class Z
Gene Aliases
CELE_Y57G7A.12
Chromosome Location
Chr.II: 1278650 - 1281039 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027312.1 NP_001022483.1 3 - 4 528 155

View Full Product Details