Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxa-4
Entrez Gene ID
4363036
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_F42G2.8
Chromosome Location
Chr.II: 2414123 - 2418048 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383799.1 8 - 9 2862 131

View Full Product Details