Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP2C18
Entrez Gene ID
415122
Gene Name
cytochrome P450 2C21
Gene Aliases
CYP2C19, CYP2C21, CYPIIC21
Chromosome Location
Chr.28: 8727010 - 8750427 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197044.1 NP_001183973.1 7 - 8 1163 85
GenBank mRNA AF049909.1 - 7 - 8 1144 85
HG965164.1 - 7 - 8 1152 85

View Full Product Details