Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLN
Entrez Gene ID
414755
Gene Name
phospholamban
Gene Aliases
-
Chromosome Location
Chr.1: 58579095 - 58590541 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003332.1 NP_001003332.1 1 - 2 82 92
GenBank mRNA Y00399.1 - - 82 92

View Full Product Details