Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD40
Entrez Gene ID
403469
Gene Name
CD40 molecule
Gene Aliases
TNFRSF5
Chromosome Location
Chr.24: 33352992 - 33383488 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002982.1 NP_001002982.1 5 - 6 692 79
XM_005634663.3 XP_005634720.1 5 - 6 589 79
XM_005634664.3 XP_005634721.1 5 - 6 589 79
XM_005634665.3 XP_005634722.1 5 - 6 589 79
XM_005634666.3 XP_005634723.1 6 - 7 828 79
XM_005634667.3 XP_005634724.1 6 - 7 806 79
XM_022408802.1 XP_022264510.1 6 - 7 823 79
XM_022408803.1 XP_022264511.1 5 - 6 702 79
XM_022408804.1 XP_022264512.1 5 - 6 468 79
XM_022408805.1 XP_022264513.1 4 - 5 410 79
GenBank mRNA AY333789.1 - 5 - 6 692 79

View Full Product Details