Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB7A
Entrez Gene ID
404007
Gene Name
RAB7A, member RAS oncogene family
Gene Aliases
RAB7
Chromosome Location
Chr.20: 2704716 - 2776518 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003316.1 NP_001003316.1 - 206 79
XM_005632015.2 XP_005632072.1 - 425 79
GenBank mRNA M35522.1 - - 206 79

View Full Product Details