Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPIFA2
Entrez Gene ID
608599
Gene Name
BPI fold containing family A member 2
Gene Aliases
-
Chromosome Location
Chr.24: 22413324 - 22423717 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022409141.1 XP_022264849.1 6 - 7 700 62

View Full Product Details