Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPS36
Entrez Gene ID
478085
Gene Name
mitochondrial ribosomal protein S36
Gene Aliases
-
Chromosome Location
Chr.2: 54302992 - 54311943 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_535262.6 XP_535262.3 3 - 4 401 129

View Full Product Details