Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AP2A2
Entrez Gene ID
475994
Gene Name
adaptor related protein complex 2 subunit alpha 2
Gene Aliases
-
Chromosome Location
Chr.18: 45269962 - 45358020 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022405507.1 XP_022261215.1 15 - 16 2128 69
XM_022405508.1 XP_022261216.1 15 - 16 2125 69
XM_022405509.1 XP_022261217.1 15 - 16 2041 69
XM_022405510.1 XP_022261218.1 15 - 16 2138 69

View Full Product Details