Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NFKBIL1
Entrez Gene ID
481711
Gene Name
NFKB inhibitor like 1
Gene Aliases
-
Chromosome Location
Chr.12: 1048393 - 1057458 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014375.1 NP_001014397.1 3 - 4 554 62
XM_005627064.3 XP_005627121.1 3 - 4 617 62
XM_005627065.3 XP_005627122.1 3 - 4 685 62

View Full Product Details