Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIB1
Entrez Gene ID
482039
Gene Name
tribbles pseudokinase 1
Gene Aliases
-
Chromosome Location
Chr.13: 23337788 - 23346304 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313808.1 NP_001300737.1 - 1361 92

View Full Product Details