Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STAP2
Entrez Gene ID
485041
Gene Name
signal transducing adaptor family member 2
Gene Aliases
-
Chromosome Location
Chr.20: 55325492 - 55336547 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022407203.1 XP_022262911.1 7 - 8 725 74
XM_022407204.1 XP_022262912.1 7 - 8 725 74
XM_022407205.1 XP_022262913.1 7 - 8 725 74
XM_542159.5 XP_542159.2 7 - 8 725 74

View Full Product Details