Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC16A5
Entrez Gene ID
483308
Gene Name
solute carrier family 16 member 5
Gene Aliases
-
Chromosome Location
Chr.9: 5362063 - 5383220 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022423897.1 XP_022279605.1 - 513 66

View Full Product Details